siRNA Details
| Gene Symbol: Tmprss11g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177162.4 | NP_796136.2 | 7 | 649 | ||
| GenBank mRNA | AK036981.1 | - | 7 | 650 | ||
| AK037029.1 | - | 3 | 147 | |||
| BC118611.1 | - | 7 | 627 | 40107184 | ||
| BC118660.1 | - | 7 | 626 | 40107187 | ||

Tmprss11g
Genomic Map