siRNA Details
| Gene Symbol: Vwc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177164.3 | NP_796138.2 | 2 | 1138 | ||
| GenBank mRNA | AB374230.1 | - | 1 | 334 | ||
| AK080585.1 | - | 4 | 587 | |||
| AK083856.1 | - | 2 | 1103 | |||
| BC064448.1 | - | 2 | 951 | |||
| BC094541.1 | - | 2 | 1138 | 6410144 | ||
| EF552208.1 | - | 1 | 402 | |||
| EF552209.1 | - | 1 | 337 | |||
| EF552210.1 | - | 2 | 196 | |||

Vwc2l
Genomic Map