siRNA Details
| Gene Symbol: Vwc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177164.3 | NP_796138.2 | 4 | 1372 | ||
| GenBank mRNA | AB374230.1 | - | 3 | 568 | ||
| AK080585.1 | - | 6 | 821 | |||
| BC064448.1 | - | 3 | 1055 | |||
| BC094541.1 | - | 4 | 1372 | 6410144 | ||
| EF552208.1 | - | 3 | 636 | |||

Vwc2l
Genomic Map