siRNA Details
| Gene Symbol: Heatr5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177171.4 | NP_796145.2 | 3 | 486 | ||
| GenBank mRNA | AK047221.1 | - | 3 | 462 | ||
| AK053055.1 | - | 3 | 487 | |||
| BC086656.1 | - | 2 | 311 | 30544145 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Heatr5a| Gene Symbol: Heatr5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177171.4 | NP_796145.2 | 3 | 486 | ||
| GenBank mRNA | AK047221.1 | - | 3 | 462 | ||
| AK053055.1 | - | 3 | 487 | |||
| BC086656.1 | - | 2 | 311 | 30544145 | ||