siRNA Details
| Gene Symbol: Ubn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177185.4 | NP_796159.3 | 14 | 2911 | ||
| GenBank mRNA | AK044162.1 | - | 18 | |||
| AK051609.1 | - | 14 | 2284 | |||
| AK173339.1 | - | 2 | 3947 | |||
| BC043095.1 | - | 2207 | ||||
| BC051458.1 | - | 14 | 2883 | 6468017 | ||

Ubn2
Genomic Map