siRNA Details
| Gene Symbol: Fam169a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100458.1 | NP_001093928.1 | 4 | 319 | ||
| NM_001146045.1 | NP_001139517.1 | 4 | 286 | |||
| GenBank mRNA | AK173051.1 | - | 4 | 319 | ||
| BC084589.1 | - | 4 | 299 | |||

Fam169a
Genomic Map