siRNA Details
Gene Symbol: Fam169a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100458.1 | NP_001093928.1 | 2, 3 | 203 | ||
NM_001146045.1 | NP_001139517.1 | 2, 3 | 170 | |||
GenBank mRNA | AK173051.1 | - | 2, 3 | 203 | ||
BC084589.1 | - | 2, 3 | 183 |
