siRNA Details
| Gene Symbol: Idi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177197.4 | NP_796171.1 | 5 | 535 | ||
| GenBank mRNA | AK029364.1 | - | 5 | 536 | ||
| AK081319.1 | - | 5 | 536 | |||
| AY263178.1 | - | 5 | 536 | |||
| BC099586.1 | - | 5 | 522 | 30292324 | ||
| BC125310.1 | - | 5 | 521 | 40129825 | ||
| BC138100.1 | - | 5 | 521 | 8861119 | ||

Idi2
Genomic Map