siRNA Details
| Gene Symbol: Tmem88b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033394.3 | NP_001028566.2 | 1 | 351 | ||
| GenBank mRNA | AK035998.1 | - | 1 | 351 | ||
| AK158459.1 | - | 1 | 352 | |||
| BC147318.1 | - | 1 | 306 | 9056362 | ||
| BC147319.1 | - | 1 | 306 | 9056365 | ||

Tmem88b
Genomic Map