siRNA Details
| Gene Symbol: Phf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113354.1 | NP_001106825.1 | 7 | 1482 | ||
| NM_177201.5 | NP_796175.1 | 7 | 1482 | |||
| GenBank mRNA | AK036609.1 | - | 7 | 856 | ||
| AK040943.1 | - | 7 | 851 | |||
| AK040969.1 | - | 7 | 1483 | |||
| AK122447.1 | - | 6 | 573 | |||
| AK134137.1 | - | 7 | 819 | |||
| AK147939.1 | - | 7 | 869 | |||
| BC138899.1 | - | 7 | 751 | 8861921 | ||

Phf8
Genomic Map