siRNA Details
| Gene Symbol: Snrnp200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177214.4 | NP_796188.2 | 37 | 5369 | ||
| GenBank mRNA | AK149291.1 | - | 8 | 892 | ||
| AK173025.1 | - | 32 | 4598 | |||
| BC011390.1 | - | 10 | 1558 | |||
| BC023685.1 | - | 10 | 1483 | |||
| BC030856.1 | - | 9 | 1310 | |||
| BC055940.1 | - | 1 | 157 | |||
| BC058608.1 | - | 6 | 852 | |||
| BC063261.1 | - | 37 | 5369 | 6408607 | ||

Snrnp200
Genomic Map