siRNA Details
| Gene Symbol: Zbtb26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199025.2 | NP_950190.2 | 2 | 1048 | ||
| GenBank mRNA | AK044592.1 | - | 2 | 679 | ||
| AK173205.1 | - | 2 | 673 | |||
| BC060655.1 | - | 2 | 1048 | 30355035 | ||
| BC109349.1 | - | 2 | 1031 | 40056733 | ||
| BC109350.1 | - | 2 | 1031 | 40056735 | ||

Zbtb26
Genomic Map