siRNA Details
| Gene Symbol: Ocrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177215.3 | NP_796189.2 | 8 | 822 | ||
| GenBank mRNA | AK035315.1 | - | 8 | 825 | ||
| AK041963.1 | - | 8 | 824 | |||
| AK044320.1 | - | 7 | 744 | |||
| AK047906.1 | - | 8 | 841 | |||
| AK138269.1 | - | 8 | 824 | |||
| BC068146.1 | - | 8 | 822 | 30533468 | ||

Ocrl
Genomic Map