siRNA Details
Gene Symbol: Cyb5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205227.1 | NP_001192156.1 | 5 | 434 | ||
NM_177216.4 | NP_796190.1 | 5 | 434 | |||
GenBank mRNA | AK085272.1 | - | 5 | 399 | ||
AK147952.1 | - | 5 | 435 | |||
BC107238.1 | - | 5 | 378 | 40052160 | ||
BC107239.1 | - | 5 | 378 | 40052161 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
