siRNA Details
| Gene Symbol: D5Ertd579e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081232.3 | NP_001074701.3 | 6 | 1703 | ||
| GenBank mRNA | AK122224.1 | - | 5 | 1214 | ||
| AK134363.1 | - | 6 | 1664 | |||
| AK139370.1 | - | 6 | 1620 | |||
| AK152987.1 | - | 6 | 1603 | |||
| BC094673.1 | - | 1 | 236 | |||
| BC151085.1 | - | 5 | 1448 | 9087998 | ||

D5Ertd579e
Genomic Map