siRNA Details
| Gene Symbol: Cass4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080820.2 | NP_001074289.1 | 5 | 1711 | ||
| NM_001276422.1 | NP_001263351.1 | 5 | 1717 | |||
| NM_001276423.1 | NP_001263352.1 | 5 | 1711 | |||
| GenBank mRNA | AK036419.1 | - | 5 | 1685 | ||
| AK080960.1 | - | 5 | 1686 | |||
| AK089450.1 | - | 5 | 1712 | |||
| AK170726.1 | - | 5 | 1711 | |||
| BC117858.1 | - | 5 | 1605 | 40095182 | ||
| BC117859.1 | - | 5 | 1604 | 40095183 | ||
| BC129975.1 | - | 5 | 1602 | 40095184 | ||
| BC129976.1 | - | 5 | 1610 | 40095185 | ||

Cass4
Genomic Map