siRNA Details
| Gene Symbol: Iffo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028273.1 | - | 1 | 238 | ||
| AK031129.1 | - | 1 | 219 | |||
| AK044382.1 | - | 1 | 240 | |||
| AK044709.1 | - | 1 | 237 | |||
| AK044728.1 | - | 1 | 230 | |||
| AK053568.1 | - | 1 | 237 | |||
| AK163571.1 | - | 1 | 239 | |||
| BY729723.1 | - | 1 | 256 | |||

Iffo1
Genomic Map