siRNA Details
| Gene Symbol: Iffo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028273.1 | - | 6 | 1307 | ||
| AK031129.1 | - | 6 | 1287 | |||
| AK044709.1 | - | 6 | 1474 | |||
| AK044728.1 | - | 7 | 1306 | |||
| AK053568.1 | - | 4 | 2694 | |||
| AK163571.1 | - | 6 | 1306 | |||
| AK169554.1 | - | 5 | 1353 | |||
| AK184406.1 | - | 2 | 138 | |||
| AK189508.1 | - | 2 | 138 | |||
| BC057601.1 | - | 7 | 752 | 6403474 | ||

Iffo1
Genomic Map