siRNA Details
| Gene Symbol: PTS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000317.2 | NP_000308.1 | 4 | 284 | ||
| GenBank mRNA | AK310768.1 | - | 3 | 238 | ||
| BC009686.1 | - | 4 | 248 | 3901665 | ||
| BC018029.1 | - | 4 | 280 | 4819386 | ||
| D17400.1 | - | 4 | 273 | |||
| M97655.1 | - | 4 | 214 | |||

PTS
Genomic Map