siRNA Details
| Gene Symbol: Ccdc158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177230.3 | NP_796204.1 | 17 | 2678 | ||
| GenBank mRNA | AK029997.1 | - | 17 | 2678 | ||
| BC137976.1 | - | 17 | 2659 | 8860995 | ||
| BC145072.1 | - | 17 | 2660 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc158| Gene Symbol: Ccdc158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177230.3 | NP_796204.1 | 17 | 2678 | ||
| GenBank mRNA | AK029997.1 | - | 17 | 2678 | ||
| BC137976.1 | - | 17 | 2659 | 8860995 | ||
| BC145072.1 | - | 17 | 2660 | |||