siRNA Details
| Gene Symbol: PTX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002852.3 | NP_002843.2 | 3 | 1168 | ||
| GenBank mRNA | AK312705.1 | - | 3 | 1168 | ||
| BC039733.1 | - | 3 | 1141 | 5768379 | ||
| M31166.1 | - | 3 | 1098 | |||
| X63053.1 | - | 3 | 1030 | |||
| X63613.1 | - | 3 | 1092 | |||

PTX3
Genomic Map