siRNA Details
| Gene Symbol: Soga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164663.1 | NP_001158135.1 | 12 | 3742 | ||
| GenBank mRNA | AK036375.1 | - | 2 | 216 | ||
| AK147227.1 | - | 12 | 3733 | |||
| AK173052.1 | - | 6 | 916 | |||
| FJ977045.2 | - | 12 | 2993 | |||

Soga1
Genomic Map