siRNA Details
| Gene Symbol: Soga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164663.1 | NP_001158135.1 | 13, 14 | 3824 | ||
| GenBank mRNA | AK036375.1 | - | 3, 4 | 298 | ||
| AK147227.1 | - | 13, 14 | 3815 | |||
| AK173052.1 | - | 7, 8 | 998 | |||
| FJ977045.2 | - | 13, 14 | 3075 | |||

Soga1
Genomic Map