siRNA Details
| Gene Symbol: Trappc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177240.3 | NP_796214.2 | 23 | 2751 | ||
| GenBank mRNA | AK083445.1 | - | 23 | 2749 | ||
| AK160812.1 | - | 22 | 3075 | |||
| BC063065.1 | - | 16 | 2361 | |||
| BC094246.1 | - | 7 | 833 | |||
| BC147504.1 | - | 23 | 2669 | 9056419 | ||
| BC151159.1 | - | 23 | 2669 | 9088048 | ||

Trappc11
Genomic Map