siRNA Details
| Gene Symbol: Pptc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177242.4 | NP_796216.2 | 2 | 565 | ||
| GenBank mRNA | AK078914.1 | - | 2 | 573 | ||
| AK083481.1 | - | 2 | 579 | |||
| AK135118.1 | - | 2 | 599 | |||
| AK162261.1 | - | 2 | 129 | |||
| BC038622.1 | - | 2 | 205 | |||
| BC068149.1 | - | 2 | 565 | 30537082 | ||

Pptc7
Genomic Map