siRNA Details
| Gene Symbol: Pptc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177242.4 | NP_796216.2 | 5 | 1122 | ||
| GenBank mRNA | AK078914.1 | - | 5 | 1130 | ||
| AK083481.1 | - | 5 | 1139 | |||
| AK135118.1 | - | 5 | 1156 | |||
| AK162261.1 | - | 5 | 686 | |||
| BC038622.1 | - | 5 | 762 | |||
| BC068149.1 | - | 5 | 1122 | 30537082 | ||

Pptc7
Genomic Map