siRNA Details
| Gene Symbol: Fastkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177244.3 | NP_796218.2 | 1 | 271 | ||
| GenBank mRNA | AK030411.1 | - | 1162 | |||
| AK083186.1 | - | 1182 | ||||
| BC075689.1 | - | 1 | 271 | 30609755 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fastkd1| Gene Symbol: Fastkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177244.3 | NP_796218.2 | 1 | 271 | ||
| GenBank mRNA | AK030411.1 | - | 1162 | |||
| AK083186.1 | - | 1182 | ||||
| BC075689.1 | - | 1 | 271 | 30609755 | ||