siRNA Details
| Gene Symbol: Tmem154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177260.2 | NP_796234.1 | 2 | 377 | ||
| GenBank mRNA | AK037111.1 | - | 2 | 289 | ||
| AK081441.1 | - | 2 | 377 | |||
| AK154236.1 | - | 2 | 292 | |||
| AK170385.1 | - | 2 | 245 | |||
| AK170626.1 | - | 2 | 192 | |||
| AK170665.1 | - | 2 | 225 | |||
| AK171510.1 | - | 2 | 360 | |||
| BC096432.1 | - | 2 | 260 | 5390573 | ||
| BC113174.1 | - | 2 | 317 | 40060792 | ||
| BC113175.1 | - | 2 | 317 | 40060793 | ||

Tmem154
Genomic Map