siRNA Details
| Gene Symbol: Tmem154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177260.2 | NP_796234.1 | 4, 5 | 549 | ||
| GenBank mRNA | AK037111.1 | - | 4, 5 | 461 | ||
| AK081441.1 | - | 4, 5 | 549 | |||
| AK154236.1 | - | 4, 5 | 464 | |||
| AK170385.1 | - | 4, 5 | 417 | |||
| AK170626.1 | - | 4, 5 | 364 | |||
| AK170665.1 | - | 4, 5 | 397 | |||
| BC096432.1 | - | 4, 5 | 432 | 5390573 | ||
| BC113174.1 | - | 4, 5 | 489 | 40060792 | ||
| BC113175.1 | - | 4, 5 | 489 | 40060793 | ||

Tmem154
Genomic Map