siRNA Details
| Gene Symbol: Zhx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177263.3 | NP_796237.2 | 3 | 690 | ||
| GenBank mRNA | AB099527.2 | - | 1 | 234 | ||
| AK029603.1 | - | 3 | 562 | |||
| AK030057.1 | - | 3 | 655 | |||
| AK143296.1 | - | 3 | 671 | |||
| AK147259.1 | - | 512 | ||||
| BC058111.1 | - | 3 | 690 | 6836697 | ||
| DQ317973.1 | - | 1 | 234 | |||

Zhx3
Genomic Map