siRNA Details
| Gene Symbol: Zhx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177263.3 | NP_796237.2 | 3 | 2380 | ||
| GenBank mRNA | AB099527.2 | - | 1 | 1924 | ||
| AK029603.1 | - | 3 | 2252 | |||
| AK030057.1 | - | 3 | 2345 | |||
| AK143296.1 | - | 3 | 2361 | |||
| AK147259.1 | - | 2200 | ||||
| BC058111.1 | - | 3 | 2380 | 6836697 | ||
| DQ317973.1 | - | 1 | 1924 | |||

Zhx3
Genomic Map