siRNA Details
| Gene Symbol: Ifitm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177265.4 | NP_796239.1 | 2, 3 | 561 | ||
| GenBank mRNA | AK045822.1 | - | 2, 3 | 562 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifitm10| Gene Symbol: Ifitm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177265.4 | NP_796239.1 | 2, 3 | 561 | ||
| GenBank mRNA | AK045822.1 | - | 2, 3 | 562 | ||