siRNA Details
| Gene Symbol: Ankrd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177268.4 | NP_796242.2 | 2 | 1229 | ||
| GenBank mRNA | AK052460.1 | - | 2 | 405 | ||
| AK085853.1 | - | 2 | 410 | |||
| AK140039.1 | - | 2 | 456 | |||
| BC021802.1 | - | 2 | 418 | |||
| BC037442.1 | - | 2 | 459 | |||
| BC116229.1 | - | 2 | 398 | 40091581 | ||
| BC116230.1 | - | 2 | 405 | 40091582 | ||

Ankrd16
Genomic Map