siRNA Details
| Gene Symbol: Samd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177271.3 | NP_796245.2 | 2 | 1431 | ||
| GenBank mRNA | AK045645.1 | - | 2 | 983 | ||
| AK087498.1 | - | 2 | 1433 | |||
| AK132444.1 | - | 2 | 1405 | |||
| BC145672.1 | - | 2 | 1205 | 40130975 | ||
| BC145674.1 | - | 2 | 1205 | 40130977 | ||

Samd5
Genomic Map