siRNA Details
Gene Symbol: Krt26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033397.5 | NP_001028569.2 | 5 | 968 | ||
GenBank mRNA | AK028591.1 | - | 5 | 968 | ||
AK028682.1 | - | 4 | 2101 | |||
AK132445.1 | - | 5 | 972 | |||
AK162718.1 | - | 5 | 970 | |||
BC116672.1 | - | 5 | 957 | 40107596 |
