siRNA Details
| Gene Symbol: Cdh18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081299.1 | NP_001074768.1 | 6 | 1314 | ||
| GenBank mRNA | AK045672.1 | - | 6 | 1466 | ||
| AK137430.1 | - | 6 | 1314 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh18| Gene Symbol: Cdh18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081299.1 | NP_001074768.1 | 6 | 1314 | ||
| GenBank mRNA | AK045672.1 | - | 6 | 1466 | ||
| AK137430.1 | - | 6 | 1314 | |||