siRNA Details
| Gene Symbol: Spata33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177279.4 | NP_796253.2 | 3 | 408 | ||
| GenBank mRNA | AK076656.1 | - | 3 | 384 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata33| Gene Symbol: Spata33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177279.4 | NP_796253.2 | 3 | 408 | ||
| GenBank mRNA | AK076656.1 | - | 3 | 384 | ||