siRNA Details
| Gene Symbol: Spata33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177279.4 | NP_796253.2 | 2 | 251 | ||
| GenBank mRNA | AK028607.1 | - | 1 | 368 | ||
| AK049895.1 | - | 1 | 331 | |||
| AK053679.1 | - | 1 | 131 | |||
| AK076656.1 | - | 2 | 227 | |||
| BC059919.1 | - | 1 | 316 | 6827099 | ||

Spata33
Genomic Map