siRNA Details
Gene Symbol: Cdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009865.2 | NP_033995.1 | 8 | 1855 | ||
GenBank mRNA | AF183946.1 | - | 8 | 1624 | ||
AK138606.1 | - | 8 | 1857 | |||
BC052056.1 | - | 8 | 1563 | 5696194 | ||
BC062962.1 | - | 8 | 1563 | 5696194 |
