siRNA Details
| Gene Symbol: Zfp605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163996.1 | NP_001157468.1 | 3, 4 | 511 | ||
| GenBank mRNA | AK043719.1 | - | 3, 4 | 512 | ||
| AK051473.1 | - | 4, 5 | 626 | |||
| BC007165.1 | - | 258 | 3489445 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp605| Gene Symbol: Zfp605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163996.1 | NP_001157468.1 | 3, 4 | 511 | ||
| GenBank mRNA | AK043719.1 | - | 3, 4 | 512 | ||
| AK051473.1 | - | 4, 5 | 626 | |||
| BC007165.1 | - | 258 | 3489445 | |||