siRNA Details
| Gene Symbol: Tnpo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177296.4 | NP_796270.2 | 11 | 1503 | ||
| GenBank mRNA | AK079444.1 | - | 1 | 2552 | ||
| AK145723.1 | - | 11 | 1520 | |||
| AK220184.1 | - | 4 | 406 | |||
| BC064646.1 | - | 11 | 1503 | 6313452 | ||
| BC075678.1 | - | 11 | 1716 | 30619729 | ||

Tnpo3
Genomic Map