siRNA Details
| Gene Symbol: Atp11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001798.2 | NP_001001798.1 | 24, 25 | 2970 | ||
| NM_001037863.1 | NP_001032952.1 | 24, 25 | 2970 | |||
| GenBank mRNA | AF156547.1 | - | 5, 6 | 515 | ||
| AK143860.1 | - | 23, 24 | 2848 | |||
| BC106087.1 | - | 24, 25 | 2949 | 30843359 | ||

Atp11c
Genomic Map