siRNA Details
| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159389.1 | NP_001152861.1 | 18 | 2574 | ||
| NM_177306.3 | NP_796280.1 | 15 | 2205 | |||
| GenBank mRNA | AK029809.1 | - | 15 | 2206 | ||
| BC132104.1 | - | 13 | 1888 | 40130381 | ||
| BC132294.1 | - | 13 | 1888 | 40130571 | ||
| BC145065.1 | - | 12 | 1770 | |||
| FJ804414.1 | - | 17 | 2422 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106388, XM_006256445, XM_006256444.

Rfx6
Genomic Map