siRNA Details
| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159389.1 | NP_001152861.1 | 17 | 2099 | ||
| NM_177306.3 | NP_796280.1 | 14 | 1730 | |||
| GenBank mRNA | AK029809.1 | - | 14 | 1731 | ||
| BC132104.1 | - | 12 | 1413 | 40130381 | ||
| BC132294.1 | - | 12 | 1413 | 40130571 | ||
| BC145065.1 | - | 11 | 1295 | |||
| FJ804414.1 | - | 16 | 1947 | |||

Rfx6
Genomic Map