siRNA Details
| Gene Symbol: Xpnpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177310.2 | NP_796284.1 | 3 | 355 | ||
| GenBank mRNA | AK042110.1 | - | 4 | 542 | ||
| AK088324.1 | - | 3 | 355 | |||
| BC137569.1 | - | 3 | 339 | 8860588 | ||
| BC144717.1 | - | 3 | 340 | 9053265 | ||

Xpnpep3
Genomic Map