siRNA Details
| Gene Symbol: Gpr183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183031.2 | NP_898852.2 | 2 | 1092 | ||
| GenBank mRNA | AK087951.1 | - | 2 | 1004 | ||
| AK138693.1 | - | 2 | 1094 | |||
| AK141593.1 | - | 2 | 1005 | |||
| AK153216.1 | - | 2 | 1005 | |||
| AK156005.1 | - | 2 | 1005 | |||
| AK156289.1 | - | 2 | 1012 | |||
| AY255606.1 | - | 1 | 180 | |||
| BC052868.1 | - | 2 | 999 | 30073287 | ||
More Information
Set Membership: |
|

Gpr183
Genomic Map