siRNA Details
| Gene Symbol: Fpr-rs6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177316.2 | NP_796290.2 | 1 | 490 | ||
| GenBank mRNA | BC104318.1 | - | 1 | 490 | 40044730 | |
| BC104319.1 | - | 1 | 490 | 40044732 | ||
More Information
Set Membership: |
|

Fpr-rs6
Genomic Map