siRNA Details
| Gene Symbol: Tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001491.1 | NP_001001491.1 | 5 | 513 | ||
| GenBank mRNA | AK206089.1 | - | 2 | 48 | ||
| AK207394.1 | - | 2 | 46 | |||
| BC023701.1 | - | 5 | 520 | 5345587 | ||
| BC023827.1 | - | 5 | 507 | 5343662 | ||
| BC032174.1 | - | 5 | 509 | |||
| BC032175.1 | - | 5 | 509 | |||
| BC070421.1 | - | 5 | 513 | 6403300 | ||

Tpm4
Genomic Map