siRNA Details
| Gene Symbol: Tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001491.1 | NP_001001491.1 | 2, 3 | 307 | ||
| GenBank mRNA | BC023701.1 | - | 2, 3 | 314 | 5345587 | |
| BC023827.1 | - | 2, 3 | 301 | 5343662 | ||
| BC032174.1 | - | 2, 3 | 304 | |||
| BC032175.1 | - | 2, 3 | 304 | |||
| BC070421.1 | - | 2, 3 | 307 | 6403300 | ||

Tpm4
Genomic Map