siRNA Details
| Gene Symbol: Ppip5k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178795.4 | NP_848910.3 | 9 | 1136 | ||
| GenBank mRNA | AF502585.1 | - | 9 | 1138 | ||
| AK046696.1 | - | 10 | 1178 | |||
| AK083140.1 | - | 9 | 1147 | |||
| AK122264.1 | - | 9 | 1102 | |||
| AK160847.1 | - | 6 | 2627 | |||
| AK162596.1 | - | 10 | 1180 | |||
| BC065138.1 | - | 9 | 1139 | |||
| BC138351.1 | - | 8 | 1034 | 8861371 | ||

Ppip5k1
Genomic Map